missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Acetylbenzo[b]thiophene 98.0+%, TCI America™
$240.46 - $959.98
Chemical Identifiers
| CAS | 22720-75-8 |
|---|---|
| Molecular Formula | C10H8OS |
| Molecular Weight (g/mol) | 176.23 |
| MDL Number | MFCD00090217 |
| InChI Key | SGSGCQGCVKWRNM-UHFFFAOYSA-N |
| Synonym | 2-acetylbenzothiophene, 2-acetylbenzo b thiophene, 1-benzo b thiophen-2-yl ethanone, 1-benzo b thiophen-2-yl-ethanone, 1-1-benzothiophen-2-yl ethanone, 1-1-benzothiophen-2-yl ethan-1-one, ethanone, 1-benzo b thienyl, 1-benzo b thiophen-2-yl ethan-1-one |
| PubChem CID | 89805 |
| IUPAC Name | 1-(1-benzothiophen-2-yl)ethan-1-one |
| SMILES | CC(=O)C1=CC2=CC=CC=C2S1 |
Chemical Identifiers
| 22720-75-8 | |
| 176.23 | |
| SGSGCQGCVKWRNM-UHFFFAOYSA-N | |
| 89805 | |
| CC(=O)C1=CC2=CC=CC=C2S1 |
| C10H8OS | |
| MFCD00090217 | |
| 2-acetylbenzothiophene, 2-acetylbenzo b thiophene, 1-benzo b thiophen-2-yl ethanone, 1-benzo b thiophen-2-yl-ethanone, 1-1-benzothiophen-2-yl ethanone, 1-1-benzothiophen-2-yl ethan-1-one, ethanone, 1-benzo b thienyl, 1-benzo b thiophen-2-yl ethan-1-one | |
| 1-(1-benzothiophen-2-yl)ethan-1-one |
Specifications
| 22720-75-8 | |
| C10H8OS | |
| 5 g | |
| 2-acetylbenzothiophene, 2-acetylbenzo b thiophene, 1-benzo b thiophen-2-yl ethanone, 1-benzo b thiophen-2-yl-ethanone, 1-1-benzothiophen-2-yl ethanone, 1-1-benzothiophen-2-yl ethan-1-one, ethanone, 1-benzo b thienyl, 1-benzo b thiophen-2-yl ethan-1-one | |
| CC(=O)C1=CC2=CC=CC=C2S1 | |
| 176.23 | |
| 176.23 | |
| Crystalline Powder |
| 89°C | |
| MFCD00090217 | |
| 3077 | |
| SGSGCQGCVKWRNM-UHFFFAOYSA-N | |
| 1-(1-benzothiophen-2-yl)ethan-1-one | |
| 89805 | |
| ≥98.0% (GC) | |
| 2-Acetylbenzo[b]thiophene |
Safety and Handling
EINECSNumber : (5)-6262
TSCA : No