accessibility menu, dialog, popup

UserName

H3COS

CAS RN 22720-75-8

IUPAC Name: 1-(1-benzothiophen-2-yl)ethan-1-one
Synonyms: 1-(1-benzothiophen-2-yl)ethanone
Molecular Weight (g/mol): 176.23
Molecular Formula: C10H8OS
InChi Key: SGSGCQGCVKWRNM-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC2=CC=CC=C2S1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)