missing translation for 'onlineSavingsMsg'
Learn More

2-Acetylbenzo[b]thiophene, 98%

CAS: 22720-75-8 | C10H8OS | 176.23 g/mol

$95.49 - $319.52

Chemical Identifiers

CAS 22720-75-8
Molecular Formula C10H8OS
Molecular Weight (g/mol) 176.23
MDL Number MFCD00090217
InChI Key SGSGCQGCVKWRNM-UHFFFAOYSA-N
Synonym 2-acetylbenzothiophene, 2-acetylbenzo b thiophene, 1-benzo b thiophen-2-yl ethanone, 1-benzo b thiophen-2-yl-ethanone, 1-1-benzothiophen-2-yl ethanone, 1-1-benzothiophen-2-yl ethan-1-one, ethanone, 1-benzo b thienyl, 1-benzo b thiophen-2-yl ethan-1-one
PubChem CID 89805
IUPAC Name 1-(1-benzothiophen-2-yl)ethan-1-one
SMILES CC(=O)C1=CC2=CC=CC=C2S1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2117303
View Documents
Thermo Scientific Chemicals
B2117303
1 g
Each for $95.49
Only null left
 
AAB2117306
View Documents
Thermo Scientific Chemicals
B2117306
5 g
Each for $319.52
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

22720-75-8
176.23
SGSGCQGCVKWRNM-UHFFFAOYSA-N
89805
CC(=O)C1=CC2=CC=CC=C2S1
C10H8OS
MFCD00090217
2-acetylbenzothiophene, 2-acetylbenzo b thiophene, 1-benzo b thiophen-2-yl ethanone, 1-benzo b thiophen-2-yl-ethanone, 1-1-benzothiophen-2-yl ethanone, 1-1-benzothiophen-2-yl ethan-1-one, ethanone, 1-benzo b thienyl, 1-benzo b thiophen-2-yl ethan-1-one
1-(1-benzothiophen-2-yl)ethan-1-one

Specifications

22720-75-8
C10H8OS
1 g
SGSGCQGCVKWRNM-UHFFFAOYSA-N
1-(1-benzothiophen-2-yl)ethan-1-one
89805
98%
86°C to 88°C
MFCD00090217
2-acetylbenzothiophene, 2-acetylbenzo b thiophene, 1-benzo b thiophen-2-yl ethanone, 1-benzo b thiophen-2-yl-ethanone, 1-1-benzothiophen-2-yl ethanone, 1-1-benzothiophen-2-yl ethan-1-one, ethanone, 1-benzo b thienyl, 1-benzo b thiophen-2-yl ethan-1-one
CC(=O)C1=CC2=CC=CC=C2S1
176.23
176.23
2-Acetylbenzo[b]thiophene
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 245-177-7

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only