missing translation for 'onlineSavingsMsg'
Learn More

2-Acetylbenzo[b]thiophene, 98%

Catalog No. AAB2117303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2117303 1 g
AAB2117306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2117303 Supplier Thermo Scientific Chemicals Supplier No. B2117303

CAS: 22720-75-8 | C10H8OS | 176.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 22720-75-8
Molecular Formula C10H8OS
Molecular Weight (g/mol) 176.23
MDL Number MFCD00090217
InChI Key SGSGCQGCVKWRNM-UHFFFAOYSA-N
Synonym 2-acetylbenzothiophene, 2-acetylbenzo b thiophene, 1-benzo b thiophen-2-yl ethanone, 1-benzo b thiophen-2-yl-ethanone, 1-1-benzothiophen-2-yl ethanone, 1-1-benzothiophen-2-yl ethan-1-one, ethanone, 1-benzo b thienyl, 1-benzo b thiophen-2-yl ethan-1-one
PubChem CID 89805
IUPAC Name 1-(1-benzothiophen-2-yl)ethan-1-one
SMILES CC(=O)C1=CC2=CC=CC=C2S1

Specifications

Melting Point 86°C to 88°C
Quantity 1 g
Formula Weight 176.23
Percent Purity 98%
Chemical Name or Material 2-Acetylbenzo[b]thiophene
EINECSNumber 245-177-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only