missing translation for 'onlineSavingsMsg'
Learn More

2-Acetyl-5-methylthiophene, 98%

Catalog No. p-7081321
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1128206 5 g
AAA1128214 25 g
AAA1128222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1128206 Supplier Thermo Scientific Chemicals Supplier No. A1128206

CAS: 13679-74-8 | C7H8OS | 140.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13679-74-8
Molecular Formula C7H8OS
Molecular Weight (g/mol) 140.20
MDL Number MFCD00014529
InChI Key YOSDTJYMDAEEAZ-UHFFFAOYSA-N
Synonym 2-acetyl-5-methylthiophene, 1-5-methylthiophen-2-yl ethanone, ketone, methyl 5-methyl-2-thienyl, 1-5-methyl-2-thienyl ethan-1-one, 2-methyl-5-acetylthiophene, ethanone, 1-5-methyl-2-thienyl, methylthienylcetone, 1-5-methyl-2-thienyl ethanone, 5-methyl-2-acetylthiophene, methyl 5-methyl-2-thienyl ketone
PubChem CID 83655
IUPAC Name 1-(5-methylthiophen-2-yl)ethanone
SMILES CC(=O)C1=CC=C(C)S1

Specifications

Melting Point 24°C to 28°C
Density 1.119
Boiling Point 65°C to 67°C (1 mmHg)
Refractive Index 1.5625
Quantity 5 g
Beilstein 110854
Sensitivity Light sensitive
Formula Weight 140.2
Percent Purity 98%
Chemical Name or Material 2-Acetyl-5-methylthiophene
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 237-181-2
RTECSNumber OB4972000
TSCA No
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only