accessibility menu, dialog, popup

UserName

H3COSCH3

CAS RN 13679-74-8

IUPAC Name: 1-(5-methylthiophen-2-yl)ethan-1-one
Synonyms: 1-(5-methylthiophen-2-yl)ethanone
Molecular Weight (g/mol): 140.2
Molecular Formula: C7H8OS
InChi Key: YOSDTJYMDAEEAZ-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C(C)S1
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)