missing translation for 'onlineSavingsMsg'
Learn More

2-Acetyl-5-methylfuran 98.0+%, TCI America™

Catalog No. A098325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A098325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A098325G Supplier TCI America Supplier No. A098325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1193-79-9
Molecular Formula C7H8O2
Molecular Weight (g/mol) 124.139
MDL Number MFCD00003243
InChI Key KEFJLCGVTHRGAH-UHFFFAOYSA-N
Synonym 2-acetyl-5-methylfuran, 5-methyl-2-acetylfuran, 1-5-methylfuran-2-yl ethanone, 1-5-methyl-2-furyl ethanone, 5-methyl-2-furylmethylketone, ethanone, 1-5-methyl-2-furanyl, 1-5-methyl-2-furanyl ethanone, 2-methyl-5-acetylfuran, 1-5-methyl-2-furyl ethan-1-one, methyl 5-methyl-2-furyl ketone
PubChem CID 14514
ChEBI CHEBI:562752
IUPAC Name 1-(5-methylfuran-2-yl)ethanone
SMILES CC1=CC=C(O1)C(=O)C

Specifications

Boiling Point 69°C
Quantity 25 g
Formula Weight 124.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Acetyl-5-methylfuran
EINECSNumber (5)-3347
RTECSNumber LT8528000
TSCA No