accessibility menu, dialog, popup

UserName

H3COOCH3

CAS RN 1193-79-9

IUPAC Name: 1-(5-methylfuran-2-yl)ethan-1-one
Synonyms: 2-acetyl-5-methylfuran 5-methyl-2-furylmethylketone
Molecular Weight (g/mol): 124.14
Molecular Formula: C7H8O2
InChi Key: KEFJLCGVTHRGAH-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C(C)O1
Boiling Point 
  • (3)
  • (1)
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)