missing translation for 'onlineSavingsMsg'
Learn More

2-Acetyl-5-methylfuran, 98+%

CAS: 1193-79-9 | C7H8O2 | 124.139 g/mol

Supplier:  Thermo Scientific Chemicals A1617214

 View more versions of this product

Catalog No. AAA1617214


Only null left
Explore available promotions

Description

Description

Glucose was transformed to various furan compounds and carboxylic acids derivatives by heat treatment of autoclave such as levulinic acid, 2-acetylfuran and 2-acetyl-5-methylfuran. using 2-acetyl-5-methylfuran or 1,5-dimethyl-2-pyrrolecarbonitrile as reactants, that the formation of the 4-arylation products. Reaction of 2-acetyl-5-methylfuran 6a with 2-aminoethanol pioduced in 79% yield the aminal.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glucose was transformed to various furan compounds and carboxylic acids derivatives by heat treatment of autoclave such as levulinic acid, 2-acetylfuran and 2-acetyl-5-methylfuran. using 2-acetyl-5-methylfuran or 1,5-dimethyl-2-pyrrolecarbonitrile as reactants, that the formation of the 4-arylation products. Reaction of 2-acetyl-5-methylfuran 6a with 2-aminoethanol pioduced in 79% yield the aminal.

Solubility
Slightly soluble in water. Soluble in alcohol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Specifications

2-Acetyl-5-methylfuran
1.068
80°C (176°F)
1.513
25 g
2-acetyl-5-methylfuran, 5-methyl-2-acetylfuran, 1-5-methylfuran-2-yl ethanone, 1-5-methyl-2-furyl ethanone, 5-methyl-2-furylmethylketone, ethanone, 1-5-methyl-2-furanyl, 1-5-methyl-2-furanyl ethanone, 2-methyl-5-acetylfuran, 1-5-methyl-2-furyl ethan-1-one, methyl 5-methyl-2-furyl ketone
KEFJLCGVTHRGAH-UHFFFAOYSA-N
1-(5-methylfuran-2-yl)ethanone
14514
124.14
1193-79-9
100°C to 101°C (25 mmHg)
C7H8O2
MFCD00003243
110853
Slightly soluble in water. Soluble in alcohol.
CC1=CC=C(O1)C(=O)C
124.139
CHEBI:562752
≥98%

Chemical Identifiers

1193-79-9
124.139
KEFJLCGVTHRGAH-UHFFFAOYSA-N
14514
1-(5-methylfuran-2-yl)ethanone
C7H8O2
MFCD00003243
2-acetyl-5-methylfuran, 5-methyl-2-acetylfuran, 1-5-methylfuran-2-yl ethanone, 1-5-methyl-2-furyl ethanone, 5-methyl-2-furylmethylketone, ethanone, 1-5-methyl-2-furanyl, 1-5-methyl-2-furanyl ethanone, 2-methyl-5-acetylfuran, 1-5-methyl-2-furyl ethan-1-one, methyl 5-methyl-2-furyl ketone
CHEBI:562752
CC1=CC=C(O1)C(=O)C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H227
Toxic if swallowed.
Combustible liquid.

P210-P264b-P270-P280-P301+P312-P330-P370+P378q-P501c

H227-H302

EINECSNumber : 214-779-1

RTECSNumber : LT8528000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only