missing translation for 'onlineSavingsMsg'
Learn More

2-Acetyl-5-methylfuran, 98+%

Catalog No. AAA1617209
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1617209 10 g
AAA1617214 25 g
AAA1617218 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1617209 Supplier Thermo Scientific Chemicals Supplier No. A1617209
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1193-79-9 | C7H8O2 | 124.139 g/mol

Glucose was transformed to various furan compounds and carboxylic acids derivatives by heat treatment of autoclave such as levulinic acid, 2-acetylfuran and 2-acetyl-5-methylfuran. using 2-acetyl-5-methylfuran or 1,5-dimethyl-2-pyrrolecarbonitrile as reactants, that the formation of the 4-arylation products. Reaction of 2-acetyl-5-methylfuran 6a with 2-aminoethanol pioduced in 79% yield the aminal.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glucose was transformed to various furan compounds and carboxylic acids derivatives by heat treatment of autoclave such as levulinic acid, 2-acetylfuran and 2-acetyl-5-methylfuran. using 2-acetyl-5-methylfuran or 1,5-dimethyl-2-pyrrolecarbonitrile as reactants, that the formation of the 4-arylation products. Reaction of 2-acetyl-5-methylfuran 6a with 2-aminoethanol pioduced in 79% yield the aminal.

Solubility
Slightly soluble in water. Soluble in alcohol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 1193-79-9
Molecular Formula C7H8O2
Molecular Weight (g/mol) 124.139
MDL Number MFCD00003243
InChI Key KEFJLCGVTHRGAH-UHFFFAOYSA-N
Synonym 2-acetyl-5-methylfuran, 5-methyl-2-acetylfuran, 1-5-methylfuran-2-yl ethanone, 1-5-methyl-2-furyl ethanone, 5-methyl-2-furylmethylketone, ethanone, 1-5-methyl-2-furanyl, 1-5-methyl-2-furanyl ethanone, 2-methyl-5-acetylfuran, 1-5-methyl-2-furyl ethan-1-one, methyl 5-methyl-2-furyl ketone
PubChem CID 14514
ChEBI CHEBI:562752
IUPAC Name 1-(5-methylfuran-2-yl)ethanone
SMILES CC1=CC=C(O1)C(=O)C

Specifications

Density 1.068
Boiling Point 100°C to 101°C (25 mmHg)
Flash Point 80°C (176°F)
Refractive Index 1.513
Quantity 10 g
Beilstein 110853
Solubility Information Slightly soluble in water. Soluble in alcohol.
Formula Weight 124.14
Percent Purity ≥98%
Chemical Name or Material 2-Acetyl-5-methylfuran
Hazard Category H227-H302
Hazard Statement GHS H Statement
H301-H227
Toxic if swallowed.
Combustible liquid.
Precautionary Statement P210-P264b-P270-P280-P301+P312-P330-P370+P378q-P501c
EINECSNumber 214-779-1
RTECSNumber LT8528000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only