missing translation for 'onlineSavingsMsg'
Learn More

2-Acetyl-4-chlorothiophene, 98+%

Catalog No. AAH3038406
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3038406 5 g
AAH3038414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3038406 Supplier Thermo Scientific Chemicals Supplier No. H3038406

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 34730-20-6 | C6H5ClOS | 160.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 34730-20-6
Molecular Formula C6H5ClOS
Molecular Weight (g/mol) 160.62
MDL Number MFCD00082791
InChI Key FKESGQASARHBDC-UHFFFAOYSA-N
PubChem CID 11105655
IUPAC Name 1-(4-chlorothiophen-2-yl)ethanone
SMILES CC(=O)C1=CC(Cl)=CS1

Specifications

Density 1.334
Quantity 5 g
UN Number UN2810
Synonym 1-4-chlorothiophen-2-yl ethanone, 2-acetyl-4-chlorothiophene, 1-4-chlorothien-2-yl ethanone, ethanone,1-4-chloro-2-thienyl, ethanone, 1-4-chloro-2-thienyl, 1-4-chlorothiophen-2-yl ethan-1-one, 1-4-chloro-2-thienyl ethanone, zlchem 1053, 4-chloro-2-acetylthiophene, ksc572s1r
Formula Weight 160.62
Percent Purity ≥98%
Chemical Name or Material 2-Acetyl-4-chlorothiophene
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only