accessibility menu, dialog, popup

UserName

H3COSCl

CAS RN 34730-20-6

IUPAC Name: 1-(4-chlorothiophen-2-yl)ethan-1-one
Synonyms: 1-(4-chlorothiophen-2-yl)ethanone
Molecular Weight (g/mol): 160.62
Molecular Formula: C6H5ClOS
InChi Key: FKESGQASARHBDC-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC(Cl)=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results