missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Acetyl-3-ethylpyrazine 98.0+%, TCI America™
$52.19 - $149.13
Chemical Identifiers
| CAS | 32974-92-8 |
|---|---|
| Molecular Formula | C8H10N2O |
| Molecular Weight (g/mol) | 150.181 |
| MDL Number | MFCD00038028 |
| InChI Key | PPJSYGVFDJEMRP-UHFFFAOYSA-N |
| Synonym | 2-acetyl-3-ethylpyrazine, 1-3-ethylpyrazin-2-yl ethanone, ethanone, 1-3-ethylpyrazinyl, 2-acetyl-3-ethyl-1,4-diazine, 1-3-ethylpyrazinyl ethan-1-one, unii-aq03n14xtl, 1-3-ethylpyrazinyl ethanone, 1-3-ethyl-2-pyrazinyl ethanone, 3-ethyl-2-acetylpyrazine, 2-acetyl-3-ethyl pyrazine |
| PubChem CID | 61918 |
| IUPAC Name | 1-(3-ethylpyrazin-2-yl)ethanone |
| SMILES | CCC1=NC=CN=C1C(=O)C |
Chemical Identifiers
| 32974-92-8 | |
| 150.181 | |
| PPJSYGVFDJEMRP-UHFFFAOYSA-N | |
| 61918 | |
| CCC1=NC=CN=C1C(=O)C |
| C8H10N2O | |
| MFCD00038028 | |
| 2-acetyl-3-ethylpyrazine, 1-3-ethylpyrazin-2-yl ethanone, ethanone, 1-3-ethylpyrazinyl, 2-acetyl-3-ethyl-1,4-diazine, 1-3-ethylpyrazinyl ethan-1-one, unii-aq03n14xtl, 1-3-ethylpyrazinyl ethanone, 1-3-ethyl-2-pyrazinyl ethanone, 3-ethyl-2-acetylpyrazine, 2-acetyl-3-ethyl pyrazine | |
| 1-(3-ethylpyrazin-2-yl)ethanone |
Specifications
| 32974-92-8 | |
| 77°C | |
| MFCD00038028 | |
| 2-acetyl-3-ethylpyrazine, 1-3-ethylpyrazin-2-yl ethanone, ethanone, 1-3-ethylpyrazinyl, 2-acetyl-3-ethyl-1,4-diazine, 1-3-ethylpyrazinyl ethan-1-one, unii-aq03n14xtl, 1-3-ethylpyrazinyl ethanone, 1-3-ethyl-2-pyrazinyl ethanone, 3-ethyl-2-acetylpyrazine, 2-acetyl-3-ethyl pyrazine | |
| CCC1=NC=CN=C1C(=O)C | |
| 150.181 | |
| 150.18 | |
| Liquid |
| Yellow | |
| C8H10N2O | |
| 1 g | |
| PPJSYGVFDJEMRP-UHFFFAOYSA-N | |
| 1-(3-ethylpyrazin-2-yl)ethanone | |
| 61918 | |
| ≥98.0% (GC) | |
| 2-Acetyl-3-ethylpyrazine |
Safety and Handling
TSCA : Yes