CAS RN 32974-92-8
CAS RN 32974-92-8
IUPAC Name:
1-(3-ethylpyrazin-2-yl)ethan-1-one
Synonyms:
1-(3-ethylpyrazin-2-yl)ethanone
Molecular Weight (g/mol):
150.18
Molecular Formula:
C8H10N2O
InChi Key:
PPJSYGVFDJEMRP-UHFFFAOYSA-N
SMILES:
CCC1=NC=CN=C1C(C)=O
1
–
1
of
1
results
1
2-Acetyl-3-ethylpyrazine 98.0+%, TCI America™
CAS: 32974-92-8 Molecular Formula: C8H10N2O Molecular Weight (g/mol): 150.181 MDL Number: MFCD00038028 InChI Key: PPJSYGVFDJEMRP-UHFFFAOYSA-N Synonym: 2-acetyl-3-ethylpyrazine,1-3-ethylpyrazin-2-yl ethanone,ethanone, 1-3-ethylpyrazinyl,2-acetyl-3-ethyl-1,4-diazine,1-3-ethylpyrazinyl ethan-1-one,unii-aq03n14xtl,1-3-ethylpyrazinyl ethanone,1-3-ethyl-2-pyrazinyl ethanone,3-ethyl-2-acetylpyrazine,2-acetyl-3-ethyl pyrazine PubChem CID: 61918 IUPAC Name: 1-(3-ethylpyrazin-2-yl)ethanone SMILES: CCC1=NC=CN=C1C(=O)C