missing translation for 'onlineSavingsMsg'
Learn More

2-Acetyl-1-naphthol, 98+%

Catalog No. p-7024592
Change view
Click to view available options
Quantity:
25 g
100 g
Catalog No. Quantity
AAA1953214 25 g
AAA1953222 100 g
2 options

Catalog No. AAA1953214

Supplier: Thermo Scientific Chemicals A1953214

Only null left

CAS: 711-79-5 | C12H10O2 | 186.21 g/mol

2-acetyl-1-naphthol and 1-acetyl-2-naphthol with triphosgene gave naphtho[1,2-e]-1,3-oxazines, naphtho[2,1-e]-1,3-oxazines or their spiro dimers depending on the molar ratio of triphosgene used for the cyclization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-acetyl-1-naphthol and 1-acetyl-2-naphthol with triphosgene gave naphtho[1,2-e]-1,3-oxazines, naphtho[2,1-e]-1,3-oxazines or their spiro dimers depending on the molar ratio of triphosgene used for the cyclization.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 711-79-5
Molecular Formula C12H10O2
Molecular Weight (g/mol) 186.21
MDL Number MFCD00003963
InChI Key JBGJVMVWYWUVOW-UHFFFAOYSA-N
Synonym 1'-hydroxy-2'-acetonaphthone, 2-acetyl-1-naphthol, 1-hydroxy-2-acetylnaphthalene, 2-acetyl-1-hydroxynaphthalene, 1-hydroxy-2-acetonaphthone, ethanone, 1-1-hydroxy-2-naphthalenyl, 1-1-hydroxynaphthalen-2-yl ethanone, 1-1-hydroxy-2-naphthalenyl ethanone, 2'-acetonaphthone, 1'-hydroxy, 1-hydroxy-2-naphthyl methyl ketone
PubChem CID 69733
IUPAC Name 1-(1-hydroxynaphthalen-2-yl)ethanone
SMILES CC(=O)C1=CC=C2C=CC=CC2=C1O

Specifications

Melting Point 98°C to 100°C
Quantity 25 g
Beilstein 1868084
Solubility Information Insoluble in water.
Formula Weight 186.21
Percent Purity 99%
Chemical Name or Material 2-Acetyl-1-naphthol
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 211-918-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only