missing translation for 'onlineSavingsMsg'
Learn More

2-Acetyl-1-naphthol, 99%

Catalog No. p-49404
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC195000050 5 g Glass bottle
AC195000250 25 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC195000050 Supplier Thermo Scientific Chemicals Supplier No. 195000050

CAS: 711-79-5 | C12H10O2 | 186.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 711-79-5
Molecular Formula C12H10O2
Molecular Weight (g/mol) 186.21
MDL Number MFCD00003963
InChI Key JBGJVMVWYWUVOW-UHFFFAOYSA-N
Synonym 1'-hydroxy-2'-acetonaphthone, 2-acetyl-1-naphthol, 1-hydroxy-2-acetylnaphthalene, 2-acetyl-1-hydroxynaphthalene, 1-hydroxy-2-acetonaphthone, ethanone, 1-1-hydroxy-2-naphthalenyl, 1-1-hydroxynaphthalen-2-yl ethanone, 1-1-hydroxy-2-naphthalenyl ethanone, 2'-acetonaphthone, 1'-hydroxy, 1-hydroxy-2-naphthyl methyl ketone
PubChem CID 69733
IUPAC Name 1-(1-hydroxynaphthalen-2-yl)ethanone
SMILES CC(=O)C1=CC=C2C=CC=CC2=C1O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 97°C to 100°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula HOC10H6COCH3
Quantity 5 g
Beilstein 08, 149
Formula Weight 186.21
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 2-Acetyl-1-naphthol, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
Harmful in contact with skin.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 211-918-8
TSCA TSCA

RUO – Research Use Only