missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Acetyl-1-methylpyrrole 98.0+%, TCI America™
Supplier: TCI America A101425ML
Specifications
| 2-Acetyl-1-methylpyrrole | |
| 202°C | |
| MFCD00003089 | |
| 2-acetyl-1-methylpyrrole, n-methyl-2-acetylpyrrole, 1-1-methyl-1h-pyrrol-2-yl ethanone, 1-1-methylpyrrol-2-yl ethanone, 1-methyl-2-acetylpyrrole, 2-acetyl-n-methylpyrrole, ethanone, 1-1-methyl-1h-pyrrol-2-yl, methyl 1-methylpyrrol-2-yl ketone, 1-1-methyl-1h-pyrrol-2-yl ethan-1-one, 2-acetyl-1-methyl pyrrole | |
| CN1C=CC=C1C(C)=O | |
| 123.16 | |
| CHEBI:59982 | |
| ≥98.0% (GC) |
| 932-16-1 | |
| C7H9NO | |
| 25 mL | |
| NZFLWVDXYUGFAV-UHFFFAOYSA-N | |
| 1-(1-methyl-1H-pyrrol-2-yl)ethan-1-one | |
| 61240 | |
| 123.16 | |
| Liquid |
Chemical Identifiers
| 932-16-1 | |
| 123.16 | |
| NZFLWVDXYUGFAV-UHFFFAOYSA-N | |
| 61240 | |
| 1-(1-methyl-1H-pyrrol-2-yl)ethan-1-one |
| C7H9NO | |
| MFCD00003089 | |
| 2-acetyl-1-methylpyrrole, n-methyl-2-acetylpyrrole, 1-1-methyl-1h-pyrrol-2-yl ethanone, 1-1-methylpyrrol-2-yl ethanone, 1-methyl-2-acetylpyrrole, 2-acetyl-n-methylpyrrole, ethanone, 1-1-methyl-1h-pyrrol-2-yl, methyl 1-methylpyrrol-2-yl ketone, 1-1-methyl-1h-pyrrol-2-yl ethan-1-one, 2-acetyl-1-methyl pyrrole | |
| CHEBI:59982 | |
| CN1C=CC=C1C(C)=O |
Safety and Handling
TSCA : Yes