missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Acetyl-1-methylpyrrole 98.0+%, TCI America™
$67.11 - $221.83
Chemical Identifiers
| CAS | 932-16-1 |
|---|---|
| Molecular Formula | C7H9NO |
| Molecular Weight (g/mol) | 123.16 |
| MDL Number | MFCD00003089 |
| InChI Key | NZFLWVDXYUGFAV-UHFFFAOYSA-N |
| Synonym | 2-acetyl-1-methylpyrrole, n-methyl-2-acetylpyrrole, 1-1-methyl-1h-pyrrol-2-yl ethanone, 1-1-methylpyrrol-2-yl ethanone, 1-methyl-2-acetylpyrrole, 2-acetyl-n-methylpyrrole, ethanone, 1-1-methyl-1h-pyrrol-2-yl, methyl 1-methylpyrrol-2-yl ketone, 1-1-methyl-1h-pyrrol-2-yl ethan-1-one, 2-acetyl-1-methyl pyrrole |
| PubChem CID | 61240 |
| ChEBI | CHEBI:59982 |
| IUPAC Name | 1-(1-methyl-1H-pyrrol-2-yl)ethan-1-one |
| SMILES | CN1C=CC=C1C(C)=O |
Chemical Identifiers
| 932-16-1 | |
| 123.16 | |
| NZFLWVDXYUGFAV-UHFFFAOYSA-N | |
| 61240 | |
| 1-(1-methyl-1H-pyrrol-2-yl)ethan-1-one |
| C7H9NO | |
| MFCD00003089 | |
| 2-acetyl-1-methylpyrrole, n-methyl-2-acetylpyrrole, 1-1-methyl-1h-pyrrol-2-yl ethanone, 1-1-methylpyrrol-2-yl ethanone, 1-methyl-2-acetylpyrrole, 2-acetyl-n-methylpyrrole, ethanone, 1-1-methyl-1h-pyrrol-2-yl, methyl 1-methylpyrrol-2-yl ketone, 1-1-methyl-1h-pyrrol-2-yl ethan-1-one, 2-acetyl-1-methyl pyrrole | |
| CHEBI:59982 | |
| CN1C=CC=C1C(C)=O |
Specifications
| 932-16-1 | |
| C7H9NO | |
| 5 mL | |
| NZFLWVDXYUGFAV-UHFFFAOYSA-N | |
| 1-(1-methyl-1H-pyrrol-2-yl)ethan-1-one | |
| 61240 | |
| 123.16 | |
| Liquid |
| 202°C | |
| MFCD00003089 | |
| 2-acetyl-1-methylpyrrole, n-methyl-2-acetylpyrrole, 1-1-methyl-1h-pyrrol-2-yl ethanone, 1-1-methylpyrrol-2-yl ethanone, 1-methyl-2-acetylpyrrole, 2-acetyl-n-methylpyrrole, ethanone, 1-1-methyl-1h-pyrrol-2-yl, methyl 1-methylpyrrol-2-yl ketone, 1-1-methyl-1h-pyrrol-2-yl ethan-1-one, 2-acetyl-1-methyl pyrrole | |
| CN1C=CC=C1C(C)=O | |
| 123.16 | |
| CHEBI:59982 | |
| ≥98.0% (GC) | |
| 2-Acetyl-1-methylpyrrole |
Safety and Handling
TSCA : Yes