missing translation for 'onlineSavingsMsg'
Learn More

2-Acetyl-1-ethylpyrrole 98.0+%, TCI America™

Catalog No. A19775G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A19775G 5 g
A197725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A19775G Supplier TCI America Supplier No. A19775G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 39741-41-8
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.182
MDL Number MFCD00191234
InChI Key HQADRFRTIALOCB-UHFFFAOYSA-N
PubChem CID 61988
IUPAC Name 1-(1-ethylpyrrol-2-yl)ethanone
SMILES CCN1C=CC=C1C(=O)C

Specifications

Boiling Point 82°C
Quantity 5 g
Formula Weight 137.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Acetyl-1-ethylpyrrole