accessibility menu, dialog, popup

UserName

CH3NH3CO

CAS RN 39741-41-8

IUPAC Name: 1-(1-ethyl-1H-pyrrol-2-yl)ethan-1-one
Synonyms: 1-(1-ethylpyrrol-2-yl)ethanone
Molecular Weight (g/mol): 137.18
Molecular Formula: C8H11NO
InChi Key: HQADRFRTIALOCB-UHFFFAOYSA-N
SMILES: CCN1C=CC=C1C(C)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results