missing translation for 'onlineSavingsMsg'
Learn More

2-Acetoxybenzonitrile 97.0+%, TCI America™

Catalog No. A11185G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A11185G 5 g
A111825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A11185G Supplier TCI America Supplier No. A11185G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5715-02-6
Molecular Formula C9H7NO2
Molecular Weight (g/mol) 161.16
MDL Number MFCD00017360
InChI Key XXLKCUTUGWSJJO-UHFFFAOYSA-N
PubChem CID 79791
IUPAC Name (2-cyanophenyl) acetate
SMILES CC(=O)OC1=CC=CC=C1C#N

Specifications

Color Yellow
Boiling Point 254°C
Quantity 5 g
Synonym 2-acetoxybenzonitrile, benzonitrile,2-acetyloxy, o-acetoxybenzonitrile, pubchem13326, 2-cyanophenyl acetate, #, acmc-1aso4, benzonitrile, 2-acetyloxy, acetic acid 2-cyanophenyl ester
Formula Weight 161.16
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Acetoxybenzonitrile