missing translation for 'onlineSavingsMsg'
Learn More

2'-Acetonaphthone 98.0+%, TCI America™

Catalog No. p-7116683
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A005325G 25 g
A0053100G 100 g
A0053500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A005325G Supplier TCI America Supplier No. A005325G
Only null left

Chemical Identifiers

CAS 93-08-3
Molecular Formula C12H10O
Molecular Weight (g/mol) 170.211
MDL Number MFCD00004108
InChI Key XSAYZAUNJMRRIR-UHFFFAOYSA-N
Synonym 2-acetylnaphthalene, 2-acetonaphthone, 2'-acetonaphthone, methyl 2-naphthyl ketone, acetonaphthone, 1-naphthalen-2-yl ethanone, 1-2-naphthyl ethanone, ethanone, 1-2-naphthalenyl, oranger cyrstals, 2-naphthyl methyl ketone
PubChem CID 7122
ChEBI CHEBI:52364
IUPAC Name 1-naphthalen-2-ylethanone
SMILES CC(=O)C1=CC2=CC=CC=C2C=C1

Specifications

Melting Point 54°C
Color White-Yellow
Boiling Point 301°C
Quantity 25 g
Formula Weight 170.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2′-Acetonaphthone
EINECSNumber (4)-0394
RTECSNumber AL2988000
TSCA Yes