missing translation for 'onlineSavingsMsg'
Learn More

2-Acetamidophenol, 97%, Thermo Scientific Chemicals

Catalog No. AC102310250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC102310250 25 g Plastic bottle
AC102311000 100 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC102310250 Supplier Thermo Scientific Chemicals Supplier No. 102310250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 614-80-2 | Molecular Formula: C8H9NO2 | Molecular Weight: 151.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 614-80-2
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.16
MDL Number MFCD00002181
InChI Key ADVGKWPZRIDURE-UHFFFAOYSA-N
Synonym 2-acetamidophenol, n-2-hydroxyphenyl acetamide, 2'-hydroxyacetanilide, o-hydroxyacetanilide, 2-acetaminophenol, 2-hydroxyacetanilide, o-acetamidophenol, o-acetylamino phenol, 2-acetylamino phenol, o-acetaminophenol
PubChem CID 11972
IUPAC Name N-(2-hydroxyphenyl)acetamide
SMILES CC(=O)NC1=CC=CC=C1O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 207°C to 210°C
Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Plastic bottle
Linear Formula CH3CONHC6H4OH
Quantity 25 g
Beilstein 13,37
Solubility Information Solubility in water: .. Other solubilities: soluble in alcohol,alkaline solutions,slightly in methanol
Formula Weight 151.16
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 2-Acetamidophenol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 210-396-9

RUO – Research Use Only