missing translation for 'onlineSavingsMsg'
Learn More

2-Acetamidoethanol 90.0+%, TCI America™

Catalog No. p-7117641
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A193525G 25 g
A1935500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A193525G Supplier TCI America Supplier No. A193525G
Only null left

Chemical Identifiers

CAS 142-26-7
Molecular Formula C4H9NO2
Molecular Weight (g/mol) 103.121
MDL Number MFCD00002836
InChI Key PVCJKHHOXFKFRP-UHFFFAOYSA-N
Synonym n-2-hydroxyethyl acetamide, n-acetylethanolamine, 2-acetamidoethanol, acetamide, n-2-hydroxyethyl, acetylcolamine, 2-acetylaminoethanol, acetamide mea, n-ethanolacetamide, n-acetyl-2-aminoethanol, hydroxyethyl acetamide
PubChem CID 8880
ChEBI CHEBI:74687
IUPAC Name N-(2-hydroxyethyl)acetamide
SMILES CC(=O)NCCO

Specifications

Color Yellow
Boiling Point 167°C
Quantity 25 g
Formula Weight 103.12
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Acetamidoethanol
RTECSNumber AC3120000
TSCA Yes