accessibility menu, dialog, popup

UserName

H3CONHOH

CAS RN 142-26-7

IUPAC Name: N-(2-hydroxyethyl)acetamide
Synonyms: acetamide mea
Molecular Weight (g/mol): 103.12
Molecular Formula: C4H9NO2
InChi Key: PVCJKHHOXFKFRP-UHFFFAOYSA-N
SMILES: CC(=O)NCCO
Boiling Point 
  • (1)
  • (4)
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (1)
  • (4)
  • (1)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (4)