missing translation for 'onlineSavingsMsg'
Learn More

2-Acetamidobenzaldehyde 98.0+%, TCI America™

Catalog No. A2926200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A2926200MG 200 mg
A29261G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A2926200MG Supplier TCI America Supplier No. A2926200MG
Only null left

Chemical Identifiers

CAS 13493-47-5
Molecular Formula C9H9NO2
Molecular Weight (g/mol) 163.18
MDL Number MFCD06739581
InChI Key OWMJAQBUFVTERI-UHFFFAOYSA-N
PubChem CID 326664
IUPAC Name N-(2-formylphenyl)acetamide
SMILES CC(=O)NC1=CC=CC=C1C=O

Specifications

Melting Point 67°C
Color Yellow
Boiling Point 103°C
Quantity 200 mg
Formula Weight 163.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Acetamidobenzaldehyde