missing translation for 'onlineSavingsMsg'
Learn More

2-Acetamido-6-nitrobenzoic Acid 98.0+%, TCI America™

Catalog No. A18415G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A18415G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A18415G Supplier TCI America Supplier No. A18415G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 73721-78-5
Molecular Formula C9H8N2O5
Molecular Weight (g/mol) 224.172
InChI Key HHNTZMHFBQIQAK-UHFFFAOYSA-N
Synonym N-Acetyl-6-nitroanthranilic Acid, 2′-Carboxy-3′-nitroacetanilide
PubChem CID 153453
IUPAC Name 2-acetamido-6-nitrobenzoic acid
SMILES CC(=O)NC1=C(C(=CC=C1)[N+](=O)[O-])C(=O)O

Specifications

Melting Point 214°C
Color Yellow
Quantity 5 g
Formula Weight 224.17
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Acetamido-6-nitrobenzoic Acid