missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Acetamido-6-methylpyridine 98.0+%, TCI America™
Supplier: TCI America A278025G
Specifications
| 2-Acetamido-6-methylpyridine | |
| 89°C | |
| C8H10N2O | |
| 25 g | |
| UEXYAZLLFZIXHN-UHFFFAOYSA-N | |
| N-(6-methylpyridin-2-yl)acetamide | |
| 220486 | |
| ≥98.0% (GC) |
| 5327-33-3 | |
| White-Yellow | |
| MFCD00091881 | |
| n-6-methylpyridin-2-yl acetamide, 2-acetamido-6-methylpyridine, n-6-methyl-2-pyridyl acetamide, 6-acetamido-2-picoline, acetamide, n-6-methyl-2-pyridinyl, 2-acetamido-6-picoline, acmc-1akw3, 2-picoline, 6-acetamido, 6-acetamido-2-methyl-pyridine, n-6-methyl-2-pyridinyl acetamide | |
| CC1=NC(=CC=C1)NC(=O)C | |
| 150.181 | |
| 150.18 | |
| Crystalline Powder |
Chemical Identifiers
| 5327-33-3 | |
| 150.181 | |
| UEXYAZLLFZIXHN-UHFFFAOYSA-N | |
| 220486 | |
| CC1=NC(=CC=C1)NC(=O)C |
| C8H10N2O | |
| MFCD00091881 | |
| n-6-methylpyridin-2-yl acetamide, 2-acetamido-6-methylpyridine, n-6-methyl-2-pyridyl acetamide, 6-acetamido-2-picoline, acetamide, n-6-methyl-2-pyridinyl, 2-acetamido-6-picoline, acmc-1akw3, 2-picoline, 6-acetamido, 6-acetamido-2-methyl-pyridine, n-6-methyl-2-pyridinyl acetamide | |
| N-(6-methylpyridin-2-yl)acetamide |
Safety and Handling
TSCA : No