missing translation for 'onlineSavingsMsg'
Learn More

2-Acetamido-6-methylpyridine 98.0+%, TCI America™

Catalog No. p-7118069
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A27805G 5 g
A278025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A27805G Supplier TCI America Supplier No. A27805G

Chemical Identifiers

CAS 5327-33-3
Molecular Formula C8H10N2O
Molecular Weight (g/mol) 150.181
MDL Number MFCD00091881
InChI Key UEXYAZLLFZIXHN-UHFFFAOYSA-N
Synonym n-6-methylpyridin-2-yl acetamide, 2-acetamido-6-methylpyridine, n-6-methyl-2-pyridyl acetamide, 6-acetamido-2-picoline, acetamide, n-6-methyl-2-pyridinyl, 2-acetamido-6-picoline, acmc-1akw3, 2-picoline, 6-acetamido, 6-acetamido-2-methyl-pyridine, n-6-methyl-2-pyridinyl acetamide
PubChem CID 220486
IUPAC Name N-(6-methylpyridin-2-yl)acetamide
SMILES CC1=NC(=CC=C1)NC(=O)C

Specifications

Melting Point 89°C
Color White-Yellow
Quantity 5 g
Formula Weight 150.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Acetamido-6-methylpyridine