missing translation for 'onlineSavingsMsg'
Learn More

2-Acetamido-5-nitropyridine 98.0+%, TCI America™

Catalog No. A28551G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A28551G 1 g
A28555G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A28551G Supplier TCI America Supplier No. A28551G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5093-64-1
Molecular Formula C7H7N3O3
Molecular Weight (g/mol) 181.151
MDL Number MFCD00833973
InChI Key XKAASKOXADTLIG-UHFFFAOYSA-N
Synonym 2-acetamido-5-nitropyridine, n-5-nitropyridin-2-yl acetamide, n-5-nitro-2-pyridyl acetamide, n-5-nitro-2-pyridinyl acetamide, acetamide, n-5-nitro-2-pyridinyl, pubchem1226, acmc-1ap26, 2-acetoamino-5-nitropyridine, ksc495i7p, 2-acetylamino-5-nitropyridine
PubChem CID 345125
IUPAC Name N-(5-nitropyridin-2-yl)acetamide
SMILES CC(=O)NC1=NC=C(C=C1)[N+](=O)[O-]

Specifications

Melting Point 200°C
Quantity 1 g
Formula Weight 181.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Acetamido-5-nitropyridine