accessibility menu, dialog, popup

UserName

CAS RN 5093-64-1

IUPAC Name: N-(5-nitropyridin-2-yl)acetamide
Synonyms: N-(5-nitropyridin-2-yl)acetamide
Molecular Weight (g/mol): 181.15
Molecular Formula: C7H7N3O3
InChi Key: XKAASKOXADTLIG-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=C(C=N1)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results