missing translation for 'onlineSavingsMsg'
Learn More

2-Acetamido-5-bromopyridine 98.0+%, TCI America™

Catalog No. p-7117869
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A23751G 1 g
A23755G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A23751G Supplier TCI America Supplier No. A23751G
Only null left

Chemical Identifiers

CAS 7169-97-3
Molecular Formula C7H7BrN2O
Molecular Weight (g/mol) 215.05
MDL Number MFCD00468968
InChI Key MJFCOXATGBYERZ-UHFFFAOYSA-N
Synonym 2-acetamido-5-bromopyridine, n-5-bromopyridin-2-yl acetamide, 2-acetylamino-5-bromopyridine, acetamide, n-5-bromo-2-pyridinyl, n-5-bromo-2-pyridyl acetamide, n-5-bromo-2-pyridinyl acetamide, n-5-bromo-pyridin-2-yl-acetamide, n-acetylamino-5-bromopyridine, pubchem5546
PubChem CID 293097
IUPAC Name N-(5-bromopyridin-2-yl)acetamide
SMILES CC(=O)NC1=NC=C(C=C1)Br

Specifications

Melting Point 176°C
Color White-Yellow
Quantity 1 g
Formula Weight 215.05
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Acetamido-5-bromopyridine