missing translation for 'onlineSavingsMsg'
Learn More

2-Acetamido-5-bromobenzoic Acid 98.0+%, TCI America™

Catalog No. A28395G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A28395G 5 g
A28391G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A28395G Supplier TCI America Supplier No. A28395G

Chemical Identifiers

CAS 38985-79-4
Molecular Formula C9H7BrNO3
Molecular Weight (g/mol) 257.06
MDL Number MFCD00040904
InChI Key QVABAFHRLMDDLM-UHFFFAOYSA-M
PubChem CID 520935
IUPAC Name 5-bromo-2-acetamidobenzoate
SMILES CC(=O)NC1=CC=C(Br)C=C1C([O-])=O

Specifications

Melting Point 218°C
Color White
Quantity 5 g
Synonym 5-bromo-2-acetamidobenzoic acid, 2-acetamino-5-bromobenzoic acid, n-acetyl-5-bromo-anthranilic acid, 2-acetylamino-5-bromobenzoic acid, n-acetyl-5-bromoanthranilic acid, 2-acetamido-5-bromobenzoic acid, benzoic acid,2-acetylamino-5-bromo, benzoic acid, 2-acetylamino-5-bromo
Formula Weight 258.07
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Acetamido-5-bromobenzoic Acid