missing translation for 'onlineSavingsMsg'
Learn More

2-Acetamido-4,6-O-benzylidene-2-deoxy-beta-D-glucopyranosyl Azide 98.0+%, TCI America™

Catalog No. A18111G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A18111G 1 g
A18115G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A18111G Supplier TCI America Supplier No. A18111G

Chemical Identifiers

CAS 168397-51-1
Molecular Formula C15H18N4O5
Molecular Weight (g/mol) 334.33
MDL Number MFCD07778221
InChI Key PEVZMZIOLKXJNJ-UHFFFAOYNA-N
PubChem CID 44653972
IUPAC Name N-{6-azido-8-hydroxy-2-phenyl-hexahydro-2H-pyrano[3,2-d][1,3]dioxin-7-yl}acetamide
SMILES CC(=O)NC1C(O)C2OC(OCC2OC1N=[N+]=[N-])C1=CC=CC=C1

Specifications

Color White
Quantity 1 g
Formula Weight 334.33
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Acetamido-4,6-O-benzylidene-2-deoxy-beta-D-glucopyranosyl Azide
TSCA No
Recommended Storage Freezer