accessibility menu, dialog, popup

UserName

CAS RN 168397-51-1

IUPAC Name: N-{6-azido-8-hydroxy-2-phenyl-hexahydro-2H-pyrano[3,2-d][1,3]dioxin-7-yl}acetamide
Synonyms: N-{6-azido-8-hydroxy-2-phenyl-hexahydro-2H-pyrano[3,2-d][1,3]dioxin-7-yl}acetamide
Molecular Weight (g/mol): 334.33
Molecular Formula: C15H18N4O5
InChi Key: PEVZMZIOLKXJNJ-UHFFFAOYNA-N
SMILES: CC(=O)NC1C(O)C2OC(OCC2OC1N=[N+]=[N-])C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results