missing translation for 'onlineSavingsMsg'
Learn More

2-[(9H-Fluoren-9-ylmethoxy)carbonylamino]isobutyric Acid 98.0+%, TCI America™

Catalog No. F08885G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F08885G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F08885G Supplier TCI America Supplier No. F08885G

Chemical Identifiers

CAS 94744-50-0
Molecular Formula C19H19NO4
Molecular Weight (g/mol) 325.36
MDL Number MFCD00151913
InChI Key HOZZVEPRYYCBTO-UHFFFAOYSA-N
Synonym fmoc-aib-oh, fmoc-alpha-methylalanine, fmoc-alpha-me-ala-oh, 2-fmoc-amino isobutyric acid, 2-9h-fluoren-9-yl methoxy carbonyl amino-2-methylpropanoic acid, alanine, n-9h-fluoren-9-ylmethoxy carbonyl-2-methyl, 2-9h-fluoren-9-ylmethoxy carbonyl amino-2-methylpropanoic acid, n-fmoc-2-aminoisobutyric acid, 2-9h-fluoren-9-ylmethoxy carbonylamino isobutyric acid
PubChem CID 2756096
IUPAC Name 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-2-methylpropanoic acid
SMILES CC(C)(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Specifications

Melting Point 182°C
Color White-Yellow
Quantity 5 g
Formula Weight 325.36
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-[(9H-Fluoren-9-ylmethoxy)carbonylamino]isobutyric Acid
TSCA No
Recommended Storage Refrigerator