missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-[(9H-Fluoren-9-ylmethoxy)carbonylamino]-1-ethanol 98.0+%, TCI America™
$107.80 - $375.41
Chemical Identifiers
| CAS | 105496-31-9 |
|---|---|
| Molecular Formula | C17H17NO3 |
| Molecular Weight (g/mol) | 283.33 |
| MDL Number | MFCD00235927 |
| InChI Key | XLIFWDZVNRWYKV-UHFFFAOYSA-N |
| Synonym | 2-(Fmoc-amino)-1-ethanol, N-Fmoc-ethanolamine |
| PubChem CID | 2817739 |
| IUPAC Name | (9H-fluoren-9-yl)methyl N-(2-hydroxyethyl)carbamate |
| SMILES | OCCNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12 |
Chemical Identifiers
| 105496-31-9 | |
| 283.33 | |
| XLIFWDZVNRWYKV-UHFFFAOYSA-N | |
| 2817739 | |
| OCCNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12 |
| C17H17NO3 | |
| MFCD00235927 | |
| 2-(Fmoc-amino)-1-ethanol, N-Fmoc-ethanolamine | |
| (9H-fluoren-9-yl)methyl N-(2-hydroxyethyl)carbamate |
Specifications
| 105496-31-9 | |
| White | |
| MFCD00235927 | |
| 2-(Fmoc-amino)-1-ethanol, N-Fmoc-ethanolamine | |
| OCCNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12 | |
| 283.33 | |
| 283.33 | |
| Crystalline Powder |
| 141°C | |
| C17H17NO3 | |
| 5 g | |
| XLIFWDZVNRWYKV-UHFFFAOYSA-N | |
| (9H-fluoren-9-yl)methyl N-(2-hydroxyethyl)carbamate | |
| 2817739 | |
| ≥98.0% (HPLC,N) | |
| 2-[(9H-Fluoren-9-ylmethoxy)carbonylamino]-1-ethanol |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator