missing translation for 'onlineSavingsMsg'
Learn More

2-[(9H-Fluoren-9-ylmethoxy)carbonylamino]-1-ethanol 98.0+%, TCI America™

Catalog No. F09125G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F09125G 5 g
F091225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F09125G Supplier TCI America Supplier No. F09125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 105496-31-9
Molecular Formula C17H17NO3
Molecular Weight (g/mol) 283.33
MDL Number MFCD00235927
InChI Key XLIFWDZVNRWYKV-UHFFFAOYSA-N
Synonym 2-(Fmoc-amino)-1-ethanol, N-Fmoc-ethanolamine
PubChem CID 2817739
IUPAC Name (9H-fluoren-9-yl)methyl N-(2-hydroxyethyl)carbamate
SMILES OCCNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Melting Point 141°C
Color White
Quantity 5 g
Formula Weight 283.33
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2-[(9H-Fluoren-9-ylmethoxy)carbonylamino]-1-ethanol
TSCA No
Recommended Storage Refrigerator