missing translation for 'onlineSavingsMsg'
Learn More

2-(9-Oxoxanthen-2-yl)propionic Acid 98.0+%, TCI America™

Catalog No. O03951G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O03951G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O03951G Supplier TCI America Supplier No. O03951G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 30087-33-3
Molecular Formula C16H12O4
Molecular Weight (g/mol) 268.268
InChI Key JUCCZCZDAPBGIV-UHFFFAOYSA-N
PubChem CID 161655
IUPAC Name 2-(9-oxoxanthen-2-yl)propanoic acid
SMILES CC(C1=CC2=C(C=C1)OC3=CC=CC=C3C2=O)C(=O)O

Specifications

Melting Point 164°C
Color White
Quantity 1 g
Formula Weight 268.27
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(9-Oxoxanthen-2-yl)propionic Acid
RTECSNumber ZD5536500
TSCA No