accessibility menu, dialog, popup

UserName

CAS RN 30087-33-3

IUPAC Name: 2-(9-oxo-9H-xanthen-2-yl)propanoic acid
Synonyms: 2-(9-oxoxanthen-2-yl)propanoic acid
Molecular Weight (g/mol): 268.27
Molecular Formula: C16H12O4
InChi Key: JUCCZCZDAPBGIV-UHFFFAOYNA-N
SMILES: CC(C(O)=O)C1=CC=C2OC3=CC=CC=C3C(=O)C2=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results