missing translation for 'onlineSavingsMsg'
Learn More

2-(9-Oxoxanthen-2-yl)propionic Acid 1,5,7-Triazabicyclo[4.4.0]dec-5-ene Salt 98.0+%, TCI America™

Catalog No. O03961G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O03961G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O03961G Supplier TCI America Supplier No. O03961G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1346753-09-0
Molecular Formula C23H25N3O4
Molecular Weight (g/mol) 407.47
InChI Key NHCZYSDZAMNWGB-UHFFFAOYSA-N
PubChem CID 91972053
IUPAC Name 1,2,3,4,6,7,8,9-octahydropyrimido[1,2-a]pyrimidin-5-ium;2-(9-oxoxanthen-2-yl)propanoate
SMILES CC(C1=CC2=C(C=C1)OC3=CC=CC=C3C2=O)C(=O)[O-].C1CNC2=[N+](C1)CCCN2

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 407.47
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(9-Oxoxanthen-2-yl)propionic Acid 1,5,7-Triazabicyclo[4.4.0]dec-5-ene Salt