CAS RN 1346753-09-0
IUPAC Name:
1H,2H,3H,4H,6H,7H,8H,9H-5λ⁵-[1,3]diazino[1,2-a]pyrimidin-5-ylium 2-(9-oxo-9H-xanthen-2-yl)propanoate
Synonyms:
1H,2H,3H,4H,6H,7H,8H,9H-5λ⁵-[1,3]diazino[1,2-a]pyrimidin-5-ylium 2-(9-oxoxanthen-2-yl)propanoate
Molecular Weight (g/mol):
407.47
Molecular Formula:
C23H25N3O4
InChi Key:
NHCZYSDZAMNWGB-UHFFFAOYNA-N
SMILES:
C1CNC2=[N+](C1)CCCN2.CC(C([O-])=O)C1=CC=C2OC3=CC=CC=C3C(=O)C2=C1
1
–
1
of
1
results
1
2-(9-Oxoxanthen-2-yl)propionic Acid 1,5,7-Triazabicyclo[4.4.0]dec-5-ene Salt 98.0+%, TCI America™
CAS: 1346753-09-0 Molecular Formula: C23H25N3O4 Molecular Weight (g/mol): 407.47 InChI Key: NHCZYSDZAMNWGB-UHFFFAOYSA-N PubChem CID: 91972053 IUPAC Name: 1,2,3,4,6,7,8,9-octahydropyrimido[1,2-a]pyrimidin-5-ium;2-(9-oxoxanthen-2-yl)propanoate SMILES: CC(C1=CC2=C(C=C1)OC3=CC=CC=C3C2=O)C(=O)[O-].C1CNC2=[N+](C1)CCCN2