Learn More
2,9-Dimethyl-1,10-phenanthroline hemihydrate, 98+%
CAS: 34302-69-7 | C28H26N4O | 434.54 g/mol
$134.33 - $1775.90
Chemical Identifiers
| CAS | 34302-69-7 |
|---|---|
| Molecular Formula | C28H26N4O |
| Molecular Weight (g/mol) | 434.54 |
| MDL Number | MFCD00149306,MFCD00004973,MFCD00149306,MFCD23140843 |
| InChI Key | IEBXFSLFDFHSRD-UHFFFAOYSA-N |
| Synonym | unii-2sdt9ev86w, 2sdt9ev86w, neocuproine hemihydrate mi, 1,10-phenanthroline, 2,9-dimethyl-, hemihydrate, 1,10-phenanthroline, 2,9-dimethyl-, hydrate 2:1, 2,9-dimethyl-1,10-phenthroline hemihydrate |
| PubChem CID | 67652146 |
| IUPAC Name | 2,9-dimethyl-1,10-phenanthroline;hydrate |
| SMILES | O.CC1=CC=C2C=CC3=CC=C(C)N=C3C2=N1.CC1=CC=C2C=CC3=CC=C(C)N=C3C2=N1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1139806
|
Thermo Scientific Chemicals
A1139806 |
5 g |
Each for $134.33
|
|
|||||
|
AAA1139814
|
Thermo Scientific Chemicals
A1139814 |
25 g |
Each for $497.82
|
|
|||||
|
AAA1139822
|
Thermo Scientific Chemicals
A1139822 |
100 g |
Each for $1,775.90
|
|
|||||
Description
2,9-Dimethyl-1,10-phenanthroline hemihydrate, is used as a reagent for the spectrophotometric determination of copper. It is used as an intermediate in medicine and chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,9-Dimethyl-1,10-phenanthroline hemihydrate, is used as a reagent for the spectrophotometric determination of copper. It is used as an intermediate in medicine and chemical research.
Solubility
It is soluble in methanol: 0.1 g/mL.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible to Strong oxidizing agents.
Chemical Identifiers
| 34302-69-7 | |
| 434.54 | |
| IEBXFSLFDFHSRD-UHFFFAOYSA-N | |
| 67652146 | |
| O.CC1=CC=C2C=CC3=CC=C(C)N=C3C2=N1.CC1=CC=C2C=CC3=CC=C(C)N=C3C2=N1 |
| C28H26N4O | |
| MFCD00149306,MFCD00004973,MFCD00149306,MFCD23140843 | |
| unii-2sdt9ev86w, 2sdt9ev86w, neocuproine hemihydrate mi, 1,10-phenanthroline, 2,9-dimethyl-, hemihydrate, 1,10-phenanthroline, 2,9-dimethyl-, hydrate 2:1, 2,9-dimethyl-1,10-phenthroline hemihydrate | |
| 2,9-dimethyl-1,10-phenanthroline;hydrate |
Specifications
| 34302-69-7 | |
| C28H26N4O | |
| 5 g | |
| 14,6449 | |
| It is soluble in methanol: 0.1g/mL. | |
| O.CC1=CC=C2C=CC3=CC=C(C)N=C3C2=N1.CC1=CC=C2C=CC3=CC=C(C)N=C3C2=N1 | |
| 434.54 | |
| 217.27 (208.26 Anhydrous) | |
| 2,9-Dimethyl-1,10-phenanthroline hemihydrate |
| 160°C to 164°C | |
| MFCD00149306,MFCD00004973,MFCD00149306,MFCD23140843 | |
| 153179 | |
| unii-2sdt9ev86w, 2sdt9ev86w, neocuproine hemihydrate mi, 1,10-phenanthroline, 2,9-dimethyl-, hemihydrate, 1,10-phenanthroline, 2,9-dimethyl-, hydrate 2:1, 2,9-dimethyl-1,10-phenthroline hemihydrate | |
| IEBXFSLFDFHSRD-UHFFFAOYSA-N | |
| 2,9-dimethyl-1,10-phenanthroline;hydrate | |
| 67652146 | |
| ≥98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c
H315-H318-H335
EINECSNumber : 207-601-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only