missing translation for 'onlineSavingsMsg'
Learn More

2,9-Dimethyl-1,10-phenanthroline hemihydrate, 98+%

CAS: 34302-69-7 | C28H26N4O | 434.54 g/mol

Supplier:  Thermo Scientific Chemicals A1139822

 View more versions of this product

Catalog No. AAA1139822


Only null left
Explore available promotions

Description

Description

2,9-Dimethyl-1,10-phenanthroline hemihydrate, is used as a reagent for the spectrophotometric determination of copper. It is used as an intermediate in medicine and chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,9-Dimethyl-1,10-phenanthroline hemihydrate, is used as a reagent for the spectrophotometric determination of copper. It is used as an intermediate in medicine and chemical research.

Solubility
It is soluble in methanol: 0.1 g/mL.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible to Strong oxidizing agents.
Specifications

Specifications

2,9-Dimethyl-1,10-phenanthroline hemihydrate
160°C to 164°C
MFCD00149306,MFCD00004973,MFCD00149306,MFCD23140843
153179
unii-2sdt9ev86w, 2sdt9ev86w, neocuproine hemihydrate mi, 1,10-phenanthroline, 2,9-dimethyl-, hemihydrate, 1,10-phenanthroline, 2,9-dimethyl-, hydrate 2:1, 2,9-dimethyl-1,10-phenthroline hemihydrate
IEBXFSLFDFHSRD-UHFFFAOYSA-N
2,9-dimethyl-1,10-phenanthroline;hydrate
67652146
≥98%
34302-69-7
C28H26N4O
100 g
14,6449
It is soluble in methanol: 0.1g/mL.
O.CC1=CC=C2C=CC3=CC=C(C)N=C3C2=N1.CC1=CC=C2C=CC3=CC=C(C)N=C3C2=N1
434.54
217.27 (208.26 Anhydrous)

Chemical Identifiers

34302-69-7
434.54
IEBXFSLFDFHSRD-UHFFFAOYSA-N
67652146
O.CC1=CC=C2C=CC3=CC=C(C)N=C3C2=N1.CC1=CC=C2C=CC3=CC=C(C)N=C3C2=N1
C28H26N4O
MFCD00149306,MFCD00004973,MFCD00149306,MFCD23140843
unii-2sdt9ev86w, 2sdt9ev86w, neocuproine hemihydrate mi, 1,10-phenanthroline, 2,9-dimethyl-, hemihydrate, 1,10-phenanthroline, 2,9-dimethyl-, hydrate 2:1, 2,9-dimethyl-1,10-phenthroline hemihydrate
2,9-dimethyl-1,10-phenanthroline;hydrate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c

H315-H318-H335

EINECSNumber : 207-601-9

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only