missing translation for 'onlineSavingsMsg'
Learn More

2,9-Dimethyl-1,10-phenanthroline, 98%

Reagent for the colorimetric determination of copper | CAS: 484-11-7 | C14H12N2 | 208.264 g/mol

Supplier:  Thermo Scientific Chemicals J6369814

 View more versions of this product

Catalog No. AAJ6369814

Only null left
Explore available promotions

Description

Description

2,9-Dimethyl-1,10-phenanthroline is used as a reagent for the colorimetric determination of copper and acts as a chelating agent. It is also used in the synthesis of arly ketones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,9-Dimethyl-1,10-phenanthroline is used as a reagent for the colorimetric determination of copper and acts as a chelating agent. It is also used in the synthesis of arly ketones.

Solubility
Slightly soluble in water and light petroleum. Soluble in methanol, ethanol, acetone, ether and benzene.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

2,9-Dimethyl-1,10-phenanthroline
159°C to 164°C
C14H12N2
25 g
14,6449
Slightly soluble in water and light petroleum; Soluble in methanol,ethanol,acetone,ether and benzene.
CC1=NC2=C(C=C1)C=CC3=C2N=C(C=C3)C
208.264
208.26
Powder
484-11-7
Brown to White
MFCD00004973
Light sensitive
neocuproine, neocuproin, 1,10-phenanthroline, 2,9-dimethyl, 2,9-dimethylphenanthroline, 2,9-dimethyl-o-phenanthroline, neo-cuproin, neocuproine hemihydrate, dmphen, unii-8e7d2sh3bv, 8e7d2sh3bv
IYRGXJIJGHOCFS-UHFFFAOYSA-N
2,9-dimethyl-1,10-phenanthroline
65237
98%

Chemical Identifiers

484-11-7
208.264
IYRGXJIJGHOCFS-UHFFFAOYSA-N
65237
CC1=NC2=C(C=C1)C=CC3=C2N=C(C=C3)C
C14H12N2
MFCD00004973
neocuproine, neocuproin, 1,10-phenanthroline, 2,9-dimethyl, 2,9-dimethylphenanthroline, 2,9-dimethyl-o-phenanthroline, neo-cuproin, neocuproine hemihydrate, dmphen, unii-8e7d2sh3bv, 8e7d2sh3bv
2,9-dimethyl-1,10-phenanthroline
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 207-601-9

RTECSNumber : SF8380000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only