missing translation for 'onlineSavingsMsg'
Learn More

2,9-Dimethyl-1,10-phenanthroline, 98%

Catalog No. p-7042768
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6369806 5 g
AAJ6369814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6369806 Supplier Thermo Scientific Chemicals Supplier No. J6369806

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Reagent for the colorimetric determination of copper | CAS: 484-11-7 | C14H12N2 | 208.264 g/mol

2,9-Dimethyl-1,10-phenanthroline is used as a reagent for the colorimetric determination of copper and acts as a chelating agent. It is also used in the synthesis of arly ketones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,9-Dimethyl-1,10-phenanthroline is used as a reagent for the colorimetric determination of copper and acts as a chelating agent. It is also used in the synthesis of arly ketones.

Solubility
Slightly soluble in water and light petroleum. Soluble in methanol, ethanol, acetone, ether and benzene.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 484-11-7
Molecular Formula C14H12N2
Molecular Weight (g/mol) 208.264
MDL Number MFCD00004973
InChI Key IYRGXJIJGHOCFS-UHFFFAOYSA-N
Synonym neocuproine, neocuproin, 1,10-phenanthroline, 2,9-dimethyl, 2,9-dimethylphenanthroline, 2,9-dimethyl-o-phenanthroline, neo-cuproin, neocuproine hemihydrate, dmphen, unii-8e7d2sh3bv, 8e7d2sh3bv
PubChem CID 65237
IUPAC Name 2,9-dimethyl-1,10-phenanthroline
SMILES CC1=NC2=C(C=C1)C=CC3=C2N=C(C=C3)C

Specifications

Melting Point 159°C to 164°C
Color Brown to White
Quantity 5 g
Sensitivity Light sensitive
Merck Index 14,6449
Solubility Information Slightly soluble in water and light petroleum; Soluble in methanol,ethanol,acetone,ether and benzene.
Formula Weight 208.26
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2,9-Dimethyl-1,10-phenanthroline
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 207-601-9
RTECSNumber SF8380000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only