Learn More
2,9-Dimethyl-1,10-phenanthroline, 98%
Reagent for the colorimetric determination of copper | CAS: 484-11-7 | C14H12N2 | 208.264 g/mol
Supplier: Thermo Scientific Chemicals J6369806
Description
2,9-Dimethyl-1,10-phenanthroline is used as a reagent for the colorimetric determination of copper and acts as a chelating agent. It is also used in the synthesis of arly ketones.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,9-Dimethyl-1,10-phenanthroline is used as a reagent for the colorimetric determination of copper and acts as a chelating agent. It is also used in the synthesis of arly ketones.
Solubility
Slightly soluble in water and light petroleum. Soluble in methanol, ethanol, acetone, ether and benzene.
Notes
Incompatible with strong oxidizing agents.
Specifications
| 2,9-Dimethyl-1,10-phenanthroline | |
| 159°C to 164°C | |
| C14H12N2 | |
| 5 g | |
| 14,6449 | |
| Slightly soluble in water and light petroleum; Soluble in methanol,ethanol,acetone,ether and benzene. | |
| CC1=NC2=C(C=C1)C=CC3=C2N=C(C=C3)C | |
| 208.264 | |
| 208.26 | |
| Powder |
| 484-11-7 | |
| Brown to White | |
| MFCD00004973 | |
| Light sensitive | |
| neocuproine, neocuproin, 1,10-phenanthroline, 2,9-dimethyl, 2,9-dimethylphenanthroline, 2,9-dimethyl-o-phenanthroline, neo-cuproin, neocuproine hemihydrate, dmphen, unii-8e7d2sh3bv, 8e7d2sh3bv | |
| IYRGXJIJGHOCFS-UHFFFAOYSA-N | |
| 2,9-dimethyl-1,10-phenanthroline | |
| 65237 | |
| 98% |
Chemical Identifiers
| 484-11-7 | |
| 208.264 | |
| IYRGXJIJGHOCFS-UHFFFAOYSA-N | |
| 65237 | |
| CC1=NC2=C(C=C1)C=CC3=C2N=C(C=C3)C |
| C14H12N2 | |
| MFCD00004973 | |
| neocuproine, neocuproin, 1,10-phenanthroline, 2,9-dimethyl, 2,9-dimethylphenanthroline, 2,9-dimethyl-o-phenanthroline, neo-cuproin, neocuproine hemihydrate, dmphen, unii-8e7d2sh3bv, 8e7d2sh3bv | |
| 2,9-dimethyl-1,10-phenanthroline |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 207-601-9
RTECSNumber : SF8380000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only