missing translation for 'onlineSavingsMsg'
Learn More

2,9-Dibutyl-1,10-phenanthroline 98.0+%, TCI America™

$162.17 - $790.37

Chemical Identifiers

CAS 85575-93-5
Molecular Formula C20H24N2
Molecular Weight (g/mol) 292.426
MDL Number MFCD02093459
InChI Key LSGGPELKXXFMGO-UHFFFAOYSA-N
PubChem CID 13384176
IUPAC Name 2,9-dibutyl-1,10-phenanthroline
SMILES CCCCC1=NC2=C(C=CC3=C2N=C(C=C3)CCCC)C=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D2565100MG
View Documents
TCI America
D2565100MG
100 mg
Each for $162.17
Only null left
 
D25651G
View Documents
TCI America
D25651G
1 g
Each for $790.37
Only null left
 
Description

Description

Specifications

Chemical Identifiers

85575-93-5
292.426
LSGGPELKXXFMGO-UHFFFAOYSA-N
2,9-dibutyl-1,10-phenanthroline
C20H24N2
MFCD02093459
13384176
CCCCC1=NC2=C(C=CC3=C2N=C(C=C3)CCCC)C=C1

Specifications

85575-93-5
White-Yellow
MFCD02093459
LSGGPELKXXFMGO-UHFFFAOYSA-N
2,9-dibutyl-1,10-phenanthroline
13384176
≥98.0% (N)
2,9-Dibutyl-1,10-phenanthroline
58°C
C20H24N2
100 mg
CCCCC1=NC2=C(C=CC3=C2N=C(C=C3)CCCC)C=C1
292.426
292.43
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'