accessibility menu, dialog, popup

UserName

CAS RN 85575-93-5

CH3NNH3C

CAS RN 85575-93-5

IUPAC Name: 2,9-dibutyl-1,10-phenanthroline
Synonyms: 2,9-dibutyl-1,10-phenanthroline
Molecular Weight (g/mol): 292.43
Molecular Formula: C20H24N2
InChi Key: LSGGPELKXXFMGO-UHFFFAOYSA-N
SMILES: CCCCC1=CC=C2C=CC3=CC=C(CCCC)N=C3C2=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results