missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(9,9'-Spirobi[fluoren]-3-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98.0+%, TCI America™
Supplier: TCI America P25621G
Specifications
| 2-(9,9′-Spirobi[fluoren]-3-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane | |
| 216°C | |
| C31H27BO2 | |
| (9,9′-Spirobi[fluoren]-3-yl)boronic Acid Pinacol Ester, 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-9,9′-spirobi[fluorene] | |
| B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)C4(C5=CC=CC=C5C6=CC=CC=C64)C7=CC=CC=C73 | |
| 442.365 | |
| 442.37 | |
| Crystalline Powder |
| 1346007-05-3 | |
| White-Yellow | |
| 1 g | |
| FCQPLZQSAKVCFH-UHFFFAOYSA-N | |
| 4,4,5,5-tetramethyl-2-(9,9'-spirobi[fluorene]-3-yl)-1,3,2-dioxaborolane | |
| 68211018 | |
| ≥98.0% (HPLC,T) |
Chemical Identifiers
| 1346007-05-3 | |
| 442.365 | |
| (9,9′-Spirobi[fluoren]-3-yl)boronic Acid Pinacol Ester, 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-9,9′-spirobi[fluorene] | |
| 4,4,5,5-tetramethyl-2-(9,9'-spirobi[fluorene]-3-yl)-1,3,2-dioxaborolane |
| C31H27BO2 | |
| FCQPLZQSAKVCFH-UHFFFAOYSA-N | |
| 68211018 | |
| B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)C4(C5=CC=CC=C5C6=CC=CC=C64)C7=CC=CC=C73 |
Safety and Handling
TSCA : No